Order Entry
Netherlands
ContactUsLinkComponent
3,3'-Diaminobenzidine (DAB) ≥97.0% (by titrimetric analysis) for biochemical research
Health
3,3'-Diaminobenzidine (DAB) ≥97.0% (by titrimetric analysis) for biochemical research
Supplier:  TCI
Certificates
undefined
Health
3,3'-Diaminobenzidine (DAB) ≥97.0% (by titrimetric analysis) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    175.0 to 179.0 deg-C
  • Peroxidase detection test:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together