Order Entry
Netherlands
ContactUsLinkComponent
1,2,3,4-Tetrahydronaphthalene ≥98.0% (by GC) for spectrophotometry
Irritant
Environment
1,2,3,4-Tetrahydronaphthalene ≥98.0% (by GC) for spectrophotometry
Catalog # TCIAT0713-250ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
Environment
1,2,3,4-Tetrahydronaphthalene ≥98.0% (by GC) for spectrophotometry
Catalog # TCIAT0713-250ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Absorbance (340nm):
    max. 0.100
  • Absorbance (350nm):
    max. 0.040
  • Absorbance (360nm):
    max. 0.020

Specification Test Results

About this item

 :
 :132.21 g/mol
Merck Index:14,09221
Boiling Pt:207°C
Melting Pt:–35.8 °C
Density:0,97 g/cm³ (20 °C)
Flash Pt:75°C
Storage Temperature:Ambient
EINECS:204-340-2
UN:3082
ADR:9,III
 :GHS07
 :GHS09
CAS Number:119-64-2
Formula:C₁₀H₁₂
MDL Number:MFCD00001733