
(±)-1,3-Butanediol ≥99.0% (by GC) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:107-88-0
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00004554
:90.12 g/mol
Merck Index:14,01567
Boiling Pt:207°C
Melting Pt:< –50 °C
Density:1,004 to 1.006 g/cm³ (20 °C)
Flash Pt:122°C
EINECS:203-529-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
- Protein impurity test (SDS-PAGE):to pass test
Specification Test Results
Frequently Bought Together