Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Litmus
Litmus
Litmus
TCIAL0062-25G
:
TCI
:
Litmus
TCIAL0062-25G
:
TCI
:
Pack type:
Glass bottle
Pk:
25 g
Seq:
3308
Appearance:
Dark blue powder to crystal
Sensitiveness:
to pass test
:
:
14,05547
:
215-739-6
:
1393-92-6
:
MFCD00131528