L(+)-Lysine dihydrochloride ≥98.0% (by titrimetric analysis)
TCIAL0131-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:+13.0 to +16.0 deg (C=2, H2O)
- Solubility in Water:almost transparency
:
:219.11 g/mol
:14,05636
:200.2 °C
:211-518-3
:657-26-1
:C₆H₁₆Cl₂N₂O₂
:MFCD00066389