
Dansyl hydrazide ≥97.0% (by titrimetric analysis) for HPLC labeling
Supplier: TCI
Certificates
About this item
CAS Number:33008-06-9
:
Formula:C₁₂H₁₅N₃O₂S
MDL Number:MFCD00003986
:265.34 g/mol
EINECS:251-337-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Potassium bromate Method):min. 97.0 %
- Effect test of HPLC derivatization:Effective as labeling agent for Heptanal
Specification Test Results
Frequently Bought Together