

Toluene-α-sulphonyl fluoride ≥98.0% (by GC) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:329-98-6
:
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424
:174.2 g/mol
Merck Index:14,07542
Boiling Pt:85°C
Melting Pt:91…92 °C
Storage Temperature:Ambient
EINECS:206-350-2
UN:2923
ADR:8,III
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:92.0 to 95.0 deg-C
- Enzyme inhibition test:to pass test
Specification Test Results
Frequently Bought Together