

Toluene-α-sulphonyl chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1939-99-7
:
Formula:C₇H₇ClO₂S
MDL Number:MFCD00007455
:190.65 g/mol
Boiling Pt:120°C
Melting Pt:90…93 °C
EINECS:217-717-1
UN:3261
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:90.0 to 94.0 deg-C
- Solubility in Toluene:very faint turbidity
Specification Test Results
Frequently Bought Together