Order Entry
Netherlands
ContactUsLinkComponent
Tetrapropylammonium hydroxide 10% in water
Corrosive
Tetrapropylammonium hydroxide 10% in water
Supplier:  TCI
Certificates
undefined
Corrosive
Tetrapropylammonium hydroxide 10% in water
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    10.0 to 11.0 w/w%

Specification Test Results

Frequently Bought Together