
Tetrapropylammonium hydroxide 10% in water
Supplier: TCI
Certificates
About this item
CAS Number:4499-86-9
:
Formula:C₁₂H₂₉NO
MDL Number:MFCD00009360
:203.37 g/mol
Storage Temperature:Refrigerator
EINECS:224-800-6
UN:3267
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
- Bromide:max. 0.10 %
Specification Test Results
Frequently Bought Together