Methyl-2-(dimethylamino)benzoate ≥97.0% (by GC, titration analysis)
: TCI
:10072-05-6
:
:C₁₀H₁₃NO₂
:MFCD00025858
:131°C
:122°C
:179.22 g/mol
:233-202-4
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Frequently Bought Together