N,N-Dimethylformamide diethyl acetal ≥95.0% (by GC) for esterification
Supplier: TCI
Certificates
About this item
CAS Number:1188-33-6
:
Formula:C₇H₁₇NO₂
MDL Number:MFCD00009227
:147.22 g/mol
Boiling Pt:130 °C (1013 hPa)
Density:0,86 g/cm³ (20 °C)
Flash Pt:22°C
Storage Temperature:Ambient
EINECS:214-707-9
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together