
Tetraethylammonium hydroxide 10% in water for ion pair chromatography
Catalog # TCIAI0363-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
- Absorbance (254nm):max. 0.05
- Bromide:max. 0.01 %
Specification Test Results
About this item
:
:147.26 g/mol
Merck Index:14,09201
EINECS:201-073-3
UN:3267
ADR:8,II
:GHS05
CAS Number:77-98-5
Formula:C₈H₂₁NO
MDL Number:MFCD00009024