


1,3-Cyclooctadiene ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:1700-10-3
:
Formula:C₈H₁₂
MDL Number:MFCD00001751
:108.18 g/mol
Boiling Pt:143°C
Melting Pt:–53 °C
Density:0,869 g/cm³ (20 °C)
Flash Pt:25°C
Storage Temperature:Refrigerator
UN:2520
ADR:3,III
:Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:216-929-1
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together