Order Entry
Netherlands
ContactUsLinkComponent
L(-)-Galactose ≥98.0% (by HPLC)
L(-)-Galactose ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
L(-)-Galactose ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC) (CAD):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    -75.0 to -82.0 deg (C=1, H2O)[included NH3 aq. (2+3)]

Specification Test Results

Related Information

Frequently Bought Together