
4-Cyanophenylboronic acid (contains varying amounts of Anhydride)
: TCI
:126747-14-6
:
:C₇H₆BNO₂
:MFCD01318968
:146.94 g/mol
:> 300 °C
:2811
:6.1,III
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 and le 114.0 %
Frequently Bought Together