3-Chlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:63503-60-6
:
Formula:C₆H₆BClO₂
MDL Number:MFCD00161354
:156.38 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 114.0 %
Specification Test Results
Related Information
Frequently Bought Together