
3,5-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAD2909-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
About this item
:
:190.82 g/mol
:GHS07
CAS Number:67492-50-6
Formula:C₆H₅BCl₂O₂
MDL Number:MFCD00051935