
2,5-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
: TCI
:85199-06-0
:
:C₈H₁₁BO₂
:MFCD01863525
:192°C
:149.98 g/mol
:GHS07
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 114.0 %
Frequently Bought Together