
2,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:193353-34-3
:
Formula:C₆H₅BF₂O₂
MDL Number:MFCD01863171
:157.91 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 113.0 %
Specification Test Results
Related Information
Frequently Bought Together