Bestelling invoeren
Netherlands
ContactUsLinkComponent
4-Nitrophenyl isocyanate ≥98.0% (by GC) (contains varying amounts of polymers)
Irritant
4-Nitrophenyl isocyanate ≥98.0% (by GC) (contains varying amounts of polymers)
Supplier:  TCI
Certificates
undefined
Irritant
4-Nitrophenyl isocyanate ≥98.0% (by GC) (contains varying amounts of polymers)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    56.0 to 60.0 deg-C

Specification Test Results

Frequently Bought Together