Order Entry
Netherlands
ContactUsLinkComponent
 
 
Dibutyl disulphide ≥98.5% (by GC)
  TCIAD1395-25ML
 :  TCI
 :  
undefined
 
Dibutyl disulphide ≥98.5% (by GC)
  TCIAD1395-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.5 %
  • Density (20deg.C):
    0.9340 to 0.9380 g/mL
  • Water:
    max. 0.03 %

 

 

 :
 :178.36 g/mol
 :226°C
 :–94 °C
 :0,9371 g/cm³ (20 °C)
 :93°C
 :Ambient
 :211-091-3
 :GHS07
 :629-45-8
 :C₈H₁₈S₂
 :MFCD00009467