


Phenyl isothiocyanate ≥99.0% (by GC) for HPLC labeling
Catalog # TCIAA5513-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
- Effect test of HPLC derivatization:Effective as labeling agent for DL-Alanine.
Specification Test Results
About this item
:
:135.19 g/mol
Merck Index:14,07297
Boiling Pt:71°C
Melting Pt:–21 °C
Density:1,13 g/cm³ (20 °C)
Flash Pt:88°C
EINECS:203-138-1
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS09
Storage Temperature:Refrigerator
CAS Number:103-72-0
Formula:C₇H₅NS
MDL Number:MFCD00004798