
(3-Chloropropyl)triethoxysilane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5089-70-3
:
Formula:ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
MDL Number:MFCD00018985
Boiling Pt:102°C
Flash Pt:86°C
:240.8 g/mol
EINECS:225-805-6
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together