1,8-Naphthosultone ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:83-31-8
:
Formula:C₁₀H₆O₃S
EINECS:201-468-0
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:156.0 to 160.0 deg-C
Specification Test Results
Frequently Bought Together