Order Entry
Netherlands
ContactUsLinkComponent
 
Dimethyl allylphosphonate tech. 85%, Technical Grade
Catalog # L10567.06
CAS Number:  
Dimethyl allylphosphonate tech. 85%, Technical Grade
Catalog # L10567.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:757-54-0
 :
Formula:C₅H₁₁O₃P
MDL Number:MFCD00014951
 :150.12 g/mol
Boiling Pt:41…42 °C (0.5 mmHg)
Storage Temperature:Ambient