



Benzylmagnesium chloride ∼16% (w/w) ∼1 mol/L in THF
Catalog # TCIAB1933-250G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):15.0 to 17.0 w/w%
Specification Test Results
About this item
:
:150.89 g/mol
Storage Temperature:Ambient
EINECS:230-039-0
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:6921-34-2
Formula:C₇H₇ClMg
MDL Number:MFCD00000469