Pesticide reference standard, Cyprodinil 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL11909000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:121552-61-2
- Description:Cyprodinil 10 µg/ml in isooctane
- Formula:C₁₄H₁₅N₃
- Description:Cyprodinil