
Dehydroabietylamine (mixture of isomeres) >55.0% (by GC)
: TCI
:99306-87-3
:1446-61-3
:C₂₀H₃₁N
:MFCD00213430
:285.47 g/mol
:215-899-7
:GHS07
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 55.0 %
- Specific rotation [a]20/D:+38.0 to +42.0 deg (C=4, MeOH)
Frequently Bought Together