


Borane dimethylamine complex ≥95.0% (by titrimetric analysis)
: TCI
:74-94-2
:
:C₂H₁₀BN
:MFCD00051068
:58.92 g/mol
:43°C
:Refrigerator
:200-823-7
:2926
:4.1,II
:GHS02
:GHS05
:GHS06
- Appearance:White powder to crystal
- Purity (Iodometric Titration):min. 95.0 %
- Melting point:35.0 to 38.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together