Borane dimethylamine complex ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 74-94-2 Molecular Formula: BH3.NH(CH3)2 Formula Weight: 58.92 Physical Form: Crystalline Density: 0.69 Appearance: White Melting Point: 36° Boiling Point: 59-65°/1-2mm UN No.: 2925 MDL No.: MFCD00051068
Synonyms: Boron hydride dimethylamine, Borane, dimethylamine complex
- CAS Number:74-94-2
- Formula:C₂H₁₀BN
- MDL Number:MFCD00051068
- Molecular Weight:58.92 g/mol
- Boiling Pt:59…65 °C (1…2 mmHg)
- Melting Pt:36 °C
- Density:0.69
- Storage Temperature:Ambient
- EINECS:200-823-7
- ADR:4.1,III
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 74-94-2 Molecular Formula: BH3.NH(CH3)2 Formula Weight: 58.92 Physical Form: Crystalline Density: 0.69 Appearance: White Melting Point: 36° Boiling Point: 59-65°/1-2mm UN No.: 2925 MDL No.: MFCD00051068
Synonyms: Boron hydride dimethylamine, Borane, dimethylamine complex
- CAS Number:74-94-2
- Formula:C₂H₁₀BN
- MDL Number:MFCD00051068
- Molecular Weight:58.92 g/mol
- Boiling Pt:59…65 °C (1…2 mmHg)
- Melting Pt:36 °C
- Density:0.69
- Storage Temperature:Ambient
- EINECS:200-823-7
- ADR:4.1,III



