1-Diazo-2-naphthol-4-sulphonic acid
TCIAD1674-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance:Light yellow to Brown powder to crystal
- Purity (Neutralization titration):min. 97.0 % (calcd.on anh.substance)
- Water:max. 15.0 %
:
:250.23 g/mol
:212-958-9
:2925
:GHS02
:GHS05
:887-76-3
:C₁₀H₆N₂O₄S
:MFCD00012409
:4.1 II