
4-(Trifluoromethylthio)benzaldehyde ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4021-50-5
:
Formula:C₈H₅F₃OS
MDL Number:MFCD00129191
:206.19 g/mol
Boiling Pt:85°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 90.0 %
Specification Test Results
Frequently Bought Together