
Diphenyl phosphite (contains varying amounts of phenol and (C₆H₅O)₃P)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4712-55-4
:
Formula:C₁₂H₁₁O₃P
MDL Number:MFCD00044497
Boiling Pt:218…219 °C (26 mmHg)
Melting Pt:12 °C
Density:1.217 g/cm³
Flash Pt:176 °C
EINECS:225-202-8
ADR:8,III
:234.19 g/mol
UN:3265
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5565 to 1.5585 (20°C, 589 nm)
- Color scale:≤50 APHA
- Specific gravity:(25/15.5°C) 1.2 to 1.22
- Acid value:≤15 mg KOH/g
- Iron (Fe):≤10 ppm
- Potassium (K):≤50 ppm
- Sodium (Na):≤100 ppm
- Insoluble matter:≤0.01 %
Specification Test Results
Frequently Bought Together