Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Progesterone 10 µg/ml in methanol
Organic reference standard, Progesterone 10 µg/ml in methanol
Catalog # PROONE9310
CAS Number:  
Organic reference standard, Progesterone 10 µg/ml in methanol
Catalog # PROONE9310
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    57-83-0
  • Description:
    Progesterone 10 µg/ml in methanol
  • Formula:
    C₂₁H₃₀O₂
  • Description:
    Progesterone

Specifications

About this item