
4,4'-Methylenebis(2,6-diethylaniline) ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13680-35-8
:
Formula:C₂₁H₃₀N₂
MDL Number:MFCD00071552
:310.4763 g/mol
Boiling Pt:205°C
EINECS:237-185-4
REACH:01-0000015192-78
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:87.0 to 91.0 deg-C
Specification Test Results
Frequently Bought Together