
2-Hydroxy-1,4-naphthoquinone ≥98.0% (by titrimetric analysis)
: TCI
:83-72-7
:
:C₁₀H₆O₃
:MFCD00001678
:174.16 g/mol
:14,05393
:201-496-3
:GHS07
- Appearance:Orange to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
Frequently Bought Together