1-Naphthylacetonitrile
Catalog # MOLEM28726321
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:167.21 g/mol
EINECS:205-078-1
UN:3439
ADR:6.1,III
CAS Number:132-75-2
Formula:C₁₂H₉N
MDL Number:MFCD00004041