
4-Chlorophenyl phosphorodichloridate ≥97.0% (by titrimetric analysis) phosphorylating agent
Supplier: TCI
Certificates
About this item
CAS Number:772-79-2
:
Formula:C₆H₄Cl₃O₂P
MDL Number:MFCD00009705
:245.43 g/mol
Boiling Pt:265°C
Flash Pt:106°C
EINECS:212-254-1
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together