
2-Chlorophenyl phosphorodichloridate ≥97.0% (by titrimetric analysis) phosphorylating agent
Catalog # TCIAC0976-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
About this item
:
:245.43 g/mol
Boiling Pt:256°C
Flash Pt:105°C
EINECS:239-129-4
UN:3265
ADR:8,III
:GHS05
CAS Number:15074-54-1
Formula:C₆H₄Cl₃O₂P
MDL Number:MFCD00009704