4-Chloro-2-(trifluoromethyl)quinoline
Catalog # MAYBKM07599.10
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:231.61 g/mol
CAS Number:1701-24-2
Formula:C₁₀H₅ClF₃N
MDL Number:MFCD00153105