

α,α,α,α',α',α'-Hexafluoro-o-xylene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:433-95-4
:
Formula:C₈H₄F₆
MDL Number:MFCD03094218
Boiling Pt:142°C
EINECS:207-092-3
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together