Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, 5-Methylchrysene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, 5-Methylchrysene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20885000AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 5-Methylchrysene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20885000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    3697-24-3
  • Description:
    5-Methylchrysene 10 µg/ml in acetonitrile
  • Formula:
    C₁₉H₁₄
  • Description:
    5-Methylchrysene

Specifications

About this item