Order Entry
Netherlands
ContactUsLinkComponent
 
Organic reference standard, 2-Methylchrysene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20870000AL
 :  
Organic reference standard, 2-Methylchrysene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20870000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    3351-32-4
  • Description:
    2-Methylchrysene 10 µg/ml in acetonitrile
  • Formula:
    C₁₉H₁₄
  • Description:
    2-Methylchrysene