p-Tolyl phenylacetate ≥99.0% (by GC)
Catalog # TCIAC0870-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:75.0 to 77.0 deg-C
Specification Test Results
About this item
:
Merck Index:14,02585
Boiling Pt:148°C
EINECS:202-990-1
CAS Number:101-94-0
Formula:C₁₅H₁₄O₂
MDL Number:MFCD00025983