Organic reference standard, n-Nonane 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15623000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:111-84-2
- Description:n-Nonane 10 µg/ml in cyclohexane
- Formula:C₉H₂₀
- Description:n-Nonane