
2-Hydroxypyridine-N-oxide ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0672-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:149.0 to 152.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
About this item
:
:111.10 g/mol
EINECS:236-100-8
:GHS07
CAS Number:13161-30-3
Formula:C₅H₅NO₂
MDL Number:MFCD00006195