

O-Methylhydroxylamine hydrochloride ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:593-56-6
:
Formula:CH₆ClNO
MDL Number:MFCD00012951
:83.515 g/mol
Merck Index:14,05989
Melting Pt:149°C
EINECS:209-798-7
UN:3261
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together