
4,4'-Bis(dimethylamino)benzhydrol ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAB0480-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:102.0 to 105.0 deg-C
Specification Test Results
About this item
:
EINECS:204-335-5
:GHS07
CAS Number:119-58-4
Formula:C₁₇H₂₂N₂O
MDL Number:MFCD00008313