Order Entry
Netherlands
ContactUsLinkComponent
Allyl mercaptan ≥70% (by GC)
Flammable
Irritant
Allyl mercaptan ≥70% (by GC)
Catalog # TCIAA0777-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Allyl mercaptan ≥70% (by GC)
Catalog # TCIAA0777-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 70.0 %

Specification Test Results

About this item

 :
 :74.15 g/mol
Boiling Pt:69°C
Flash Pt:21°C
Storage Temperature:Refrigerator
EINECS:212-792-7
UN:3336
ADR:3,II
 :GHS02
 :GHS07
CAS Number:870-23-5
Formula:C₃H₆S
MDL Number:MFCD00004894